MilliporeSigma
  • CENP-A nucleosome clusters form rosette-like structures around HJURP during G1.
CENP-A nucleosome clusters form rosette-like structures around HJURP during G1.
Nature communications (2019-10-02)
Leonid Andronov, Khalid Ouararhni, Isabelle Stoll, Bruno P Klaholz, Ali Hamiche
ABSTRACT
CENP-A is an essential histone H3 variant that epigenetically marks the centromeric region of chromosomes. Here we show that CENP-A nucleosomes form characteristic clusters during the G1 phase of the cell cycle. 2D and 3D super-resolution microscopy and segmentation analysis reveal that these clusters encompass a globular rosette-like structure, which evolves into a more compact structure in late G1. The rosette-like clusters contain numerous CENP-A molecules and form a large cellular structure of ∼250-300 nm diameter with remarkably similar shapes for each centromere. Co-localization analysis shows that HJURP, the CENP-A chaperone, is located in the center of the rosette and serves as a nucleation point. The discovery of an HJURP-mediated CENP-A nucleation in human cells and its structural description provide important insights into the mechanism of CENP-A deposition and the organization of CENP-A chromatin in the centromeric region.
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Cysteamine, ~95%
Sigma-Aldrich
Cysteamine, ≥98.0% (RT)
Sigma-Aldrich
2,2′-Thiodiethanol, ≥99%
Sigma-Aldrich
Sodium DL-lactate solution, syrup, 60 % (w/w), synthetic
Sigma-Aldrich
Cysteamine hydrochloride, ≥97.0% (RT)