MilliporeSigma
  • New Amine and Aromatic Substituted Analogues of Phencyclidine: Synthesis and Determination of Acute and Chronic Pain Activities.
New Amine and Aromatic Substituted Analogues of Phencyclidine: Synthesis and Determination of Acute and Chronic Pain Activities.
Combinatorial chemistry & high throughput screening (2019-10-18)
Maryam Shokrollahi, Marjaneh Samadizadeh, Mohsen Khalili, Seyed A Sobhanian, Abbas Ahmadi
ABSTRACT
Phencyclidine (PCP, I) is a synthetic drug with remarkable physiological properties. PCP and its analogues exert many pharmacological activities and interact with some neurotransmitter systems in the central nervous system like particular affinity for PCP sites in NMDA receptors or dopamine uptake blocking or even both. The following research, methyl group with electron-donating and dipole moment characters was added in different positions of phenyl ring along with the substitution of benzylamine (with many pharmacological effects) instead of piperidine ring of I to produce new compounds (II-V) of this family with more analgesic activities. Analgesic activities of these new compounds were measured by tail immersion and formalin tests for acute and chronic pains, respectively. Also, the outcomes were compared with control and PCP (10 mg/kg) groups. The results indicate that compounds III, IV, and V have more acute and chronic antinociceptive effects than PCP and compound II which may be concerned with more antagonizing activities of these new painkillers for the blockage of dopamine reuptake as well as high affinity for NMDA receptors PCP binding site. It can be concluded that the benzylamine derivative of phencyclidine with a methyl group on the benzyl position on phenyl ring (V) is a more appropriate candidate to reduce acute and chronic (thermal and chemical) pains compared to other substituted phenyl analogs (II-IV) and PCP.
Sigma-Aldrich
Benzyl bromide, reagent grade, 98%
Sigma-Aldrich
2-Bromotoluene, 99%