MilliporeSigma
  • Bilobalide Suppresses Adipogenesis in 3T3-L1 Adipocytes via the AMPK Signaling Pathway.
Bilobalide Suppresses Adipogenesis in 3T3-L1 Adipocytes via the AMPK Signaling Pathway.
Molecules (Basel, Switzerland) (2019-10-02)
Su Bu, Chun Ying Yuan, Quan Xue, Ying Chen, Fuliang Cao
ABSTRACT
Bilobalide, the only sesquiterpene compound from Ginkgo biloba leaf, exhibits various beneficial pharmaceutical activities, such as antioxidant, anti-inflammation, and protective effects for the central nervous system. Several bioactive components extracted from Ginkgo biloba extract reportedly have the potential to attenuate lipid metabolism. However, the effect of bilobalide on lipid metabolism remains unclear. In this study, we used 3T3-L1 cells as the cell model to investigate the effect of bilobalide on adipogenesis. The results showed that bilobalide inhibited 3T3-L1 preadipocyte differentiation and intracellular lipid accumulation. Quantitative real-time PCR and western blotting results indicated that several specific adipogenic transcription factors and a few important adipogenesis-related genes were significantly down regulated on both mRNA and protein levels in bilobalide treatment groups. By contrast, the expression of some lipolytic genes, such as adipose triglyceride lipase, hormone-sensitive lipase (HSL), and carnitine palmitoyltransferase-1α, were all up-regulated by bilobalide treatment, and the phosphorylation of AMP-activated protein kinase (AMPK), acetyl-CoA carboxylase 1, and HSL were stimulated. Furthermore, bilobalide treatment partially restored AMPK activity following its blockade by compound C (dorsomorphin). These results suggested that bilobalide inhibited adipogenesis and promoted lipolysis in 3T3-L1 cells by activating the AMPK signaling pathway.