MilliporeSigma
  • The effect of finishing diet supplemented with methionine/lysine and methionine/α-tocopherol on performance, carcass traits and meat quality of Hanwoo steers.
The effect of finishing diet supplemented with methionine/lysine and methionine/α-tocopherol on performance, carcass traits and meat quality of Hanwoo steers.
Asian-Australasian journal of animal sciences (2019-09-05)
Farouq Heidar Barido, Dicky Tri Utama, Hae Seong Jeong, Juntae Kim, Chang Woo Lee, Yeon Soo Park, Sung Ki Lee
ABSTRACT
This study aimed to compare the effects of diets with and without supplements of methionine/lysine (met/lys) and methionine/α-tocopherol (met/α-tocopherol) on the performance, carcass traits and meat quality of Hanwoo steers. A total of 21 Hanwoo steers were divided into three different groups. Each group consisted of 7 animals that received different dietary treatments for 120 days as follows: a control (C) diet composed of a basal diet with 74% total digestible nutrient and 12% crude protein; treatment 1 (T1) composed of a basal diet enriched with methionine 69%+lysine 31%; and treatment (T2) composed of a basal diet enriched with methionine 84.65%+α-tocopherol 15.35%. Daily supplementation was given using the top-dressing method (20 g/animal). The met/lys groups yielded a longissimus lumborum with increased water holding capacity (p<0.01) and lower shear force value (p<0.05). Dietary met/lys did not have an adverse effect on the animal performance and carcass traits. Instead, these supplements contributed significantly to the higher protein content compared to the control diet (p<0.05). Myristic acid (C14:0) was the only fatty acid affected by the supplementation. While the met/α-tocopherol group led to significantly higher protein and oxymyoglobin contents during storage (p<0.05). It also contributed significantly to the lower shear force value and 2-thiobarbituric acid reactive substances score during storage (p<0.05) compared to the control and treatment 1 since the initial storage day. The met/α-tocopherol diet also yielded meat with a redder color (p<0.05) after 3 days of storage. However, it did not significantly contribute to the fatty acid profiles of Hanwoo steers. Met/lys supplementation resulted in higher protein scores, water holding capacity and lower shear force scores. While met/α-tocopherol supplementation contributes to the production of redder meat, reduces lipid oxidation, production of more tender meat and increases the content of protein and oxymyoglobin percentage.
Product Description
Supelco
PUFA No.2 (Animal Source), analytical standard