Merck
  • Palmitic acid induces guanylin gene expression through the Toll-like receptor 4/nuclear factor-κB pathway in rat macrophages.
Palmitic acid induces guanylin gene expression through the Toll-like receptor 4/nuclear factor-κB pathway in rat macrophages.
American journal of physiology. Cell physiology (2019-09-26)
Sayaka Akieda-Asai, Hao Ma, Yukari Date
ABSTRACT
Recently, we showed that double-transgenic rats overexpressing guanylin (Gn), a bioactive peptide, and its receptor, guanylyl cyclase-C (GC-C), specifically in macrophages demonstrate an antiobesity phenotype and low-expression levels of proinflammatory cytokines in the mesenteric fat even when fed a high-fat diet. Here, we examined the levels and mechanism of Gn and GC-C transcription following saturated fatty acid and lipopolysaccharide (LPS), an activator of Toll-like receptor 4 (TLR4), exposure by using the NR8383 macrophage cell line. In addition, the levels of guanylin and cGMP were increased by addition of either palmitic acid or LPS. Next, we investigated the interaction of the gene transcription and nuclear factor-κB (NF-κB) by using an NF-κB inhibitor and chromatin immunoprecipitation assay. We showed that palmitic acid induced Gn gene expression via TLR4 and NF-κB. Moreover, we demonstrated that NF-κB binding to the Gn promoter was responsible for the induction of gene transcription by palmitic acid or LPS. Our results indicate that saturated fatty acids such as palmitic acid activate Gn gene expression via the NF-κB pathway, raising the possibility that the activated Gn-GC-C system may contribute to the inhibition of high-fat diet-induced proinflammatory cytokines in macrophages.
Sigma-Aldrich
Guanosine 3′,5′-cyclic monophosphate, ≥98% (HPLC), powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-GUCY2C (981-995) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution