MilliporeSigma
  • Do ovarian steroid hormones control the resumption of embryonic growth following the period of diapause in roe deer (Capreolus capreolus)?
Do ovarian steroid hormones control the resumption of embryonic growth following the period of diapause in roe deer (Capreolus capreolus)?
Reproductive biology (2019-05-31)
Barbara Drews, Alba Rudolf Vegas, Vera Anna van der Weijden, Vladimir Milojevic, Anna Katharina Hankele, Gerhard Schuler, Susane Ernestine Ulbrich
ABSTRACT
Embryonic diapause in the European roe deer includes a period of five months from August to December in which embryonic development is extremely decelerated. Following exit from diapause, the embryo rapidly elongates and subsequently implants. In diapausing carnivores and marsupials, resumption of embryonic growth is regulated by ovarian steroid hormones. In the roe deer, the role of steroid hormones is not known to date. In the present study, progesterone (P4), estradiol-17β (E2) and total estrogens (Etot) were determined in blood plasma and endometrium of roe deer shot in the course of regular huntings between September and December. Steroid hormone concentrations were correlated to the corresponding size of the embryo derived from ex vivo uterine flushing and to the date of sampling. The mean plasma concentrations of P4 (5.4 ± 0.2 ng/ml, mean ± SE, N = 87), E2 (24.3 ± 2.6 pg/ml, N = 86) and Etot (21.7 ± 2.6 pg/ml, N = 78) remained constant over the sampling period and were not correlated to embryonic size. Likewise, endometrial concentrations of P4 (66.1 ± 6.5 ng/g), E2 (284.0 ± 24.43 pg/g) and, Etot (440.9 ± 24.43 pg/g) showed no changes over time [corrected]. Therefore, it was concluded that ovarian steroid hormones do not play a determining role in resumption of embryonic growth following the period of diapause in the roe deer.
Sigma-Aldrich
Butyl methyl ether, 99%