MilliporeSigma
  • IgG and IgM antibodies to the refolded MOG(1-125) extracellular domain in humans.
IgG and IgM antibodies to the refolded MOG(1-125) extracellular domain in humans.
Journal of neuroimmunology (2011-01-11)
Francesca Gori, Barbara Mulinacci, Lara Massai, Carlo Avolio, Mariantonietta Caragnano, Elisa Peroni, Silvia Lori, Mario Chelli, Anna Maria Papini, Paolo Rovero, Francesco Lolli
ABSTRACT
Antibodies to MOG in serum have a dubious prognostic value in multiple sclerosis. The MOG recombinant protein conformational properties relevant to the antigenic activity are unknown. We employed a solid-phase ELISA based on a product (rMOG(ED)(His)(6)) expressed in E. coli after subcloning the cDNA of the extracellular domain of rat MOG, performing a refolding procedure on column and affinity purification. The far-UV Circular Dichroism (CD) spectra of rMOG(ED)(His)(6) showed a β-sheet, a characteristic feature of the Ig-fold. However, in MS sera and controls we failed to detected IgM or IgG antibodies.