MilliporeSigma
  • SLAMF1 is required for TLR4-mediated TRAM-TRIF-dependent signaling in human macrophages.
SLAMF1 is required for TLR4-mediated TRAM-TRIF-dependent signaling in human macrophages.
The Journal of cell biology (2018-02-15)
Maria Yurchenko, Astrid Skjesol, Liv Ryan, Gabriel Mary Richard, Richard Kumaran Kandasamy, Ninghai Wang, Cox Terhorst, Harald Husebye, Terje Espevik
ABSTRACT
Signaling lymphocytic activation molecule family 1 (SLAMF1) is an Ig-like receptor and a costimulatory molecule that initiates signal transduction networks in a variety of immune cells. In this study, we report that SLAMF1 is required for Toll-like receptor 4 (TLR4)-mediated induction of interferon β (IFNβ) and for killing of Gram-negative bacteria by human macrophages. We found that SLAMF1 controls trafficking of the Toll receptor-associated molecule (TRAM) from the endocytic recycling compartment (ERC) to Escherichia coli phagosomes. In resting macrophages, SLAMF1 is localized to ERC, but upon addition of E. coli, it is trafficked together with TRAM from ERC to E. coli phagosomes in a Rab11-dependent manner. We found that endogenous SLAMF1 protein interacted with TRAM and defined key interaction domains as amino acids 68 to 95 of TRAM as well as 15 C-terminal amino acids of SLAMF1. Interestingly, the SLAMF1-TRAM interaction was observed for human but not mouse proteins. Overall, our observations suggest that SLAMF1 is a new target for modulation of TLR4-TRAM-TRIF inflammatory signaling in human cells.
Sigma-Aldrich
4G10® Platinum, Anti-Phosphotyrosine Antibody, Biotin Conjugate, clone 4G10®, Upstate®, from mouse
Sigma-Aldrich
GeneJuice® Transfection Reagent, Non-lipid based chemical transfection reagent optimized for maximum transfection efficiency, ease-of-use, and minimal cytotoxicity on a wide variety of mammalian cells.
Millipore
Glutathione High Capacity Magnetic Agarose Beads, (1:1 suspension in a 30% ethanol solution)
Sigma-Aldrich
Anti-Glutathione-S-transferase (GST) antibody, Mouse monoclonal, clone 2H3-D10, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Interferon-α/β Receptor Chain 2 Antibody, clone MMHAR-2, clone MMHAR-2, Chemicon®, from mouse