MilliporeSigma
  • Phytochemical, in-vitro biological and chemo-preventive profiling of Arisaema jacquemontii Blume tuber extracts.
Phytochemical, in-vitro biological and chemo-preventive profiling of Arisaema jacquemontii Blume tuber extracts.
BMC complementary and alternative medicine (2019-09-16)
Saira Tabassum, Muhammad Zia, Esperanza J Carcahe de Blanco, Riffat Batool, Roohi Aslam, Sajid Hussain, Qamar Wali, Muhammad Mudassar Gulzar
ABSTRACT
Arisaema jacquemontii is traditionally used in treatment of different diseases. In this study, phytochemical, in vitro biological and chemo-preventive screening of A. jacquemontii was carried out to explore its pharmacological potential. The dried tuber of A. jacquemontii was extracted in 11 organic solvent mixture of different polarity. The extracts were screened for phytochemical assays (phenolics and flavonoids), antioxidants potential (free radical scavenging activity, total antioxidant activity, reducing power), biological activities (antibacterial, antifungal, cytotoxic, antileishmanial, protein kinase inhibition), and chemopreventive activities using different cell lines through standard protocols. Significant amount phenolic contents were determined in EtOH and MeOH extracts (210.3 ± 3.05 and 193.2 ± 3.15 μg GAE/mg, respectively). Maximum flavonoid content was determined in MeOH extract (22.4 ± 4.04 μg QE/mg). Noteworthy, DPPH scavenging activity was also recorded for MeOH extract (87.66%) followed by MeOH+EtOAc extract (85.11%). Considerable antioxidant capacity (7.8 ± 0.12 μg AAE/mg) and reducing power (3.1 ± 0.15 μg AAE/mg) was observed in extract of MeOH. The LC50 against brine shrimp and leishmanial parasite was found 9.01 and 12.87 μg/mL for n-Hex and CHCl3 extracts, respectively. The highest zone of inhibition against Streptomyces hyphae formation (12.5 ± 1.77 mm) by n-Hex extract. Growth zone of inhibition 13.8 ± 1.08 mm was recorded for EtOAc and MeOH extracts, respectively against Micrococcus luteus while 10.0 ± 0.11 mm for MeOH extract against Aspergillus flavus. In-vitro cytotoxic assay showed that n-Hex extract had higher cytotoxicity against DU-145 prostate cancer and HL-60 cancer cell lines. NF-kB and MTP potential showed 34.01 and 44.87 μg/mL for n-Hex and CHCl3 extracts, respectively in chemo-preventive potential. The study concludes that Arisaema jacquemontii bears significant phytochemical activity and pharmacological activities, this plant can be further explored for isolation of active component against a number of aliments.
Sigma-Aldrich
Terbinafine hydrochloride
Sigma-Aldrich
Paclitaxel, from Taxus yannanensis, powder
Sigma-Aldrich
Amphotericin B solution, 250 μg/mL in deionized water, 0.1 μm filtered, BioReagent, suitable for cell culture
Roxithromycin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid