Merck
  • Genomic deletion of TLR2 induces aggravated white matter damage and deteriorated neurobehavioral functions in mouse models of Alzheimer's disease.
Genomic deletion of TLR2 induces aggravated white matter damage and deteriorated neurobehavioral functions in mouse models of Alzheimer's disease.
Aging (2019-09-12)
Chao Zhou, Xiaoyu Sun, Yuting Hu, Jiaxing Song, Shuyu Dong, Delian Kong, Yuqiao Wang, Xiaodong Hua, Jingjing Han, Yan Zhou, Guoliang Jin, Xinxin Yang, Hongjuan Shi, Zuohui Zhang, Fang Hua
ABSTRACT
Toll-like receptor-2 (TLR2), a member of the TLR family, plays an important role in the initiation and regulation of immune/inflammation response, which is a critical mechanism underlying Alzheimer's disease (AD). To clarify the role of TLR2 in the pathological process of AD, in the present study, TLR2 knockout plus APPswe/PSEN1dE9 transgenic mice (AD-TLR2KO) were generated. Neurobehavioral tests and brain MRI scan were conducted on mice at the age of 12 months. Additionally, neuron loss was evaluated using NeuN staining. Amyloid β protein (Aβ), glial fibrillary acidic protein (GFAP), endogenous ligands for TLR2, and the activation of downstream signaling of TLR2 in mouse brains were detected by immunohistochemistry and Western blots. The results demonstrated that TLR2 deficit induced learning disabilities, decreased spontaneous activity, increased anxiety and depression, and led to white matter damage (WMD), brain atrophy, loss of neurons, and glial activation. Moreover, TLR2 deficit aggravated impaired neurobehavioral functions and WMD in AD mice, but did not affect the Aβ deposition in mouse brains. Our data indicate that the genomic deletion of TLR2 impairs neurobehavioral functions, induces WMD and brain atrophy, and increases the activation of astrocytes, which in turn aggravate the symptoms of AD through a non-Aβ mechanism.
Sigma-Aldrich
Monoclonal Anti-MyD88 antibody produced in mouse, clone 2E9C2, purified immunoglobulin, buffered aqueous solution