MilliporeSigma
  • TLR4 counteracts BVRA signaling in human leukocytes via differential regulation of AMPK, mTORC1 and mTORC2.
TLR4 counteracts BVRA signaling in human leukocytes via differential regulation of AMPK, mTORC1 and mTORC2.
Scientific reports (2019-05-09)
Zhiyong Zhang, Louis F Amorosa, Anna Petrova, Susette Coyle, Marie Macor, Mohan Nair, Leonard Y Lee, Beatrice Haimovich
ABSTRACT
TLR4 is implicated in diseases associated with chronic low-grade inflammation, yet homeostatic signaling mechanisms that prevent and/or are affected by chronic TLR4 activation are largely uncharacterized. We recently reported that LPS/TLR4 activates in human leukocytes signaling intermediates (SI), abbreviated TLR4-SI, which include mTORC1-specific effectors and targets, and that leukocytes of patients with T2D or after cardiopulmonary bypass (CPB) expressed similar SI. Extending these previous findings, here we show that TLR4-SI expression post-CPB was associated with low serum bilirubin and reduced preoperative expression of biliverdin reductase A (BVRA), the enzyme that converts biliverdin to bilirubin, in patient's leukocytes. Biliverdin inhibited TLR4 signaling in leukocytes and triggered phosphorylation of mTORC2-specific targets, including Akt, PKCζ, AMPKα-LKB1-TSC1/2, and their association with BVRA. Torin, PP242, and a PKCζ inhibitory peptide, but not rapamycin, prevented these biliverdin-induced responses and TLR4 inhibition. In contrast, LPS/TLR4 triggered decreases in BVRA, AMPKα and PKCζ expression, and an increase in haptoglobin, a heme binding protein, in leukocytes in vivo and in vitro, indicating that activated TLR4 may suppress biliverdin/BVRA signaling. Significantly, compared to non-diabetics, BVRA and PKCζ expression was low and haptoglobin was high in T2D patients leukocytes. Sustained TLR4 activation may deregulate homeostatic anti-inflammatory BVRA/mTORC2 signaling and thereby contribute to chronic inflammatory diseases.
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution