MilliporeSigma
  • Brd4 and P300 Confer Transcriptional Competency during Zygotic Genome Activation.
Brd4 and P300 Confer Transcriptional Competency during Zygotic Genome Activation.
Developmental cell (2019-06-19)
Shun Hang Chan, Yin Tang, Liyun Miao, Hiba Darwich-Codore, Charles E Vejnar, Jean-Denis Beaudoin, Damir Musaev, Juan P Fernandez, Maria D J Benitez, Ariel A Bazzini, Miguel A Moreno-Mateos, Antonio J Giraldez
ABSTRACT
The awakening of the genome after fertilization is a cornerstone of animal development. However, the mechanisms that activate the silent genome after fertilization are poorly understood. Here, we show that transcriptional competency is regulated by Brd4- and P300-dependent histone acetylation in zebrafish. Live imaging of transcription revealed that genome activation, beginning at the miR-430 locus, is gradual and stochastic. We show that genome activation does not require slowdown of the cell cycle and is regulated through the translation of maternally inherited mRNAs. Among these, the enhancer regulators P300 and Brd4 can prematurely activate transcription and restore transcriptional competency when maternal mRNA translation is blocked, whereas inhibition of histone acetylation blocks genome activation. We conclude that P300 and Brd4 are sufficient to trigger genome-wide transcriptional competency by regulating histone acetylation on the first zygotic genes in zebrafish. This mechanism is critical for initiating zygotic development and developmental reprogramming.
Sigma-Aldrich
Anti-Mouse IgG - Atto 594 antibody produced in goat, ~1 mg/mL protein, affinity isolated antibody
Sigma-Aldrich
Anti-Rabbit-IgG - Atto 647N antibody produced in goat, 1 mg/mL IgG
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, (H+L) HRP conjugate, 1 mg/mL, Chemicon®