MilliporeSigma
  • IGF1 Treatment Improves Cardiac Remodeling after Infarction by Targeting Myeloid Cells.
IGF1 Treatment Improves Cardiac Remodeling after Infarction by Targeting Myeloid Cells.
Molecular therapy : the journal of the American Society of Gene Therapy (2018-12-12)
Andre Heinen, Rianne Nederlof, Priyadarshini Panjwani, André Spychala, Tengis Tschaidse, Heiko Reffelt, Johannes Boy, Annika Raupach, Stefanie Gödecke, Patrick Petzsch, Karl Köhrer, Maria Grandoch, Anne Petz, Jens W Fischer, Christina Alter, Jelena Vasilevska, Philipp Lang, Axel Gödecke
ABSTRACT
Insulin-like growth factor 1 (IGF1) is an anabolic hormone that controls the growth and metabolism of many cell types. However, IGF1 also mediates cardio-protective effects after acute myocardial infarction (AMI), but the underlying mechanisms and cellular targets are not fully understood. Here we demonstrate that short-term IGF1 treatment for 3 days after AMI improved cardiac function after 1 and 4 weeks. Regional wall motion was improved in ischemic segments, scar size was reduced, and capillary density increased in the infarcted area and the border zone. Unexpectedly, inducible inactivation of the IGF1 receptor (IGF1R) in cardiomyocytes did not attenuate the protective effect of IGF1. Sequential cardiac transcriptomic analysis indicated an altered myeloid cell response in the acute phase after AMI, and, notably, myeloid-cell Igf1r-/- mice lost the protective IGF1 function after I/R. In addition, IGF1 induced an M2-like anti-inflammatory phenotype in bone marrow-derived macrophages and enhanced the number of anti-inflammatory macrophages in heart tissue on day 3 after AMI in vivo. In summary, modulation of the acute inflammatory phase after AMI by IGF1 represents an effective mechanism to preserve cardiac function after I/R.