MilliporeSigma
  • The functional and inflammatory response of brain endothelial cells to Toll-Like Receptor agonists.
The functional and inflammatory response of brain endothelial cells to Toll-Like Receptor agonists.
Scientific reports (2018-07-06)
Rebecca H Johnson, Dan T Kho, Simon J O' Carroll, Catherine E Angel, E Scott Graham
ABSTRACT
Toll-Like receptors (TLRs) represent an important early warning mechanism for the immune system to detect infection or tissue damage. The focus of this research was to determine the neuroinflammatory responses to commercial TLR ligands and their effects on brain endothelial barrier strength. Using biosensor technology we screened TLR ligands to all human TLRs and found that the brain endothelial hCMVECs cell line only responded to Poly(I:C) (TLR3-ligand), LPS (TLR4-ligand) and Imiquimod (TLR7 ligand). Both Poly(I:C) and LPS induced pronounced pro-inflammatory cytokine secretion as expected, whereas Imiquimod did not induce secretion of any pro-inflammatory cytokines. Using ECIS technology to measure endothelial barrier function, LPS and Poly(I:C) both acutely reduced barrier-strength, whereas Imiquimod caused immediate and sustained strengthening of the barrier. Further cytokine and ECIS studies showed that Imiquimod could abrogate some of the pro-inflammatory responses to Poly(I:C) and LPS. Most surprisingly, PCR revealed that the hCMVECs lacked TLR7 but expressed both TLR3 and TLR4 and did not respond to other structurally different TLR7 ligands. These data demonstrate that brain endothelial cells can be regulated by TLR 3 and TLR4 ligands in a pro-inflammatory manner and have receptors to Imiquimod, distinct to the classical TLR7, that function in an anti-inflammatory manner.
Product Description
Roche
Transcriptor Reverse Transcriptase
Sigma-Aldrich
CL097, ≥98% (HPLC)