MilliporeSigma
  • Isolation and characterization of vasohibin-2 as a homologue of VEGF-inducible endothelium-derived angiogenesis inhibitor vasohibin.
Isolation and characterization of vasohibin-2 as a homologue of VEGF-inducible endothelium-derived angiogenesis inhibitor vasohibin.
Arteriosclerosis, thrombosis, and vascular biology (2006-03-11)
Takumi Shibuya, Kazuhide Watanabe, Hiroshi Yamashita, Kazue Shimizu, Hiroki Miyashita, Mayumi Abe, Takuya Moriya, Hideki Ohta, Hikaru Sonoda, Tooru Shimosegawa, Koichi Tabayashi, Yasufumi Sato
ABSTRACT
We recently isolated vasohibin, a novel vascular endothelial growth factor (VEGF)-inducible endothelium-derived angiogenesis inhibitor. Our aim is to find DNA sequences homologous to vasohibin and determine their expression profile. By the search of DNA sequences in the database, we found one homologous gene and designated it vasohibin-2. Overall amino acid sequence homology between the prototype vasohibin (vasohibin-1) and vasohibin-2 was >50%. Vasohibin-2 exhibited antiangiogenic activity. Vasohibin-2 expression in cultured endothelial cells was low and not inducible by the stimulation that induced vasohibin-1. However, the immunohistochemical analysis revealed that vasohibin-1 and -2 were diffusely expressed in endothelial cells in embryonic organs during mid-gestation. After that time point, vasohibin-1 and -2 became faint, but persisted to a certain extent in arterial endothelial cells from late gestation to neonate. Expression of vasohibin-1 and -2 could be augmented in vivo by local transfection with the VEGF gene in the embryonic brain or by cutaneous wounding in adult mice. These results suggest that vasohibin-2, in combination with vasohibin-1, forms a novel family of angiogenesis inhibitors.