MilliporeSigma
  • SHARPIN is a component of the NF-κB-activating linear ubiquitin chain assembly complex.
SHARPIN is a component of the NF-κB-activating linear ubiquitin chain assembly complex.
Nature (2011-04-02)
Fuminori Tokunaga, Tomoko Nakagawa, Masaki Nakahara, Yasushi Saeki, Masami Taniguchi, Shin-ichi Sakata, Keiji Tanaka, Hiroyasu Nakano, Kazuhiro Iwai
ABSTRACT
Cpdm (chronic proliferative dermatitis) mice develop chronic dermatitis and an immunodeficiency with increased serum IgM, symptoms that resemble those of patients with X-linked hyper-IgM syndrome and hypohydrotic ectodermal dysplasia (XHM-ED), which is caused by mutations in NEMO (NF-κB essential modulator; also known as IKBKG). Spontaneous null mutations in the Sharpin (SHANK-associated RH domain interacting protein in postsynaptic density) gene are responsible for the cpdm phenotype in mice. SHARPIN shows significant similarity to HOIL-1L (also known as RBCK1), a component of linear ubiquitin chain assembly complex (LUBAC), which induces NF-κB activation through conjugation of linear polyubiquitin chains to NEMO. Here, we identify SHARPIN as an additional component of LUBAC. SHARPIN-containing complexes can linearly ubiquitinate NEMO and activated NF-κB. Thus, we re-define LUBAC as a complex containing SHARPIN, HOIL-1L, and HOIP (also known as RNF31). Deletion of SHARPIN drastically reduced the amount of LUBAC, which resulted in attenuated TNF-α- and CD40-mediated activation of NF-κB in mouse embryonic fibroblasts (MEFs) or B cells from cpdm mice. Considering the pleomorphic phenotype of cpdm mice, these results confirm the predicted role of LUBAC-mediated linear polyubiquitination in NF-κB activation induced by various stimuli, and strongly suggest the involvement of LUBAC-induced NF-κB activation in various disorders.