MilliporeSigma
  • ZNF521 Has an Inhibitory Effect on the Adipogenic Differentiation of Human Adipose-Derived Mesenchymal Stem Cells.
ZNF521 Has an Inhibitory Effect on the Adipogenic Differentiation of Human Adipose-Derived Mesenchymal Stem Cells.
Stem cell reviews (2018-06-26)
Emanuela Chiarella, Annamaria Aloisio, Bruna Codispoti, Giovanna Nappo, Stefania Scicchitano, Valeria Lucchino, Ylenia Montalcini, Augusto Camarotti, Olimpio Galasso, Manfredi Greco, Giorgio Gasparini, Maria Mesuraca, Heather Mandy Bond, Giovanni Morrone
ABSTRACT
Mesenchymal stem cells (MSCs) are multipotent progenitors present in the bone marrow stroma and in subcutaneous abdominal fat, an abundant and easily accessible source of MSCs with the ability to differentiate along multiple lineage pathways. The stem cell-associated transcription co-factor Zinc Finger Protein 521 (ZNF521/zfp521) has been implicated in the control of the homeostasis of hematopoietic, neural and osteo-adipogenic progenitors. Here we document through the analysis of a panel of human adipose-derived stem cells (hADSCs), that ZNF521 strongly inhibits the generation of mature adipocytes. Enforced overexpression of ZNF521 in these cells resulted in a significant delay and reduction in adipocyte differentiation upon exposure to inducers of adipogenesis. Of particular relevance, ZNF521 was able to inhibit the expression of ZNF423, recently identified as an essential commitment factor necessary for the generation of pre-adipocytes. Conversely, silencing of ZNF521 was found to significantly enhance the adipogenic differentiation of hADSCs. Inhibition of adipogenesis by ZNF521 was at least in part due to inhibition of EBF1. Taken together, these results confirm a role for ZNF521 as a key negative regulator of adipocyte differentiation of hADSCs.
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Oil Red O solution, 0.5% in isopropanol
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution