MilliporeSigma
  • The non-canonical NF-κB pathway promotes NPC2 expression and regulates intracellular cholesterol trafficking.
The non-canonical NF-κB pathway promotes NPC2 expression and regulates intracellular cholesterol trafficking.
Science China. Life sciences (2018-08-10)
Yacheng Liao, Jian Wei, Juqiong Wang, Xiongjie Shi, Jie Luo, Bao-Liang Song
ABSTRACT
Niemann-Pick type C2 (NPC2) is a lysosome luminal protein that functions in concert with NPC1 to mediate egress of low-density lipoprotein-derived cholesterol from lysosome. The nuclear factor kappa B subunit 2 (NF-κB2) protein is a component of NF-κB transcription factor complex critically implicated in immune and inflammatory responses. Here, we report that NF-κB2 regulates intracellular cholesterol transport by controlling NPC2 expression. RNAi-mediated disruption of NF-κB2, as well as other signaling members of the non-canonical NF-κB pathway, caused intracellular cholesterol accumulation. Blockage of the non-canonical NF-κB pathway suppressed NPC2 expression, whereas Lymphotoxin β receptor (LTβR) activation or Baff receptor (BaffR) stimulation up-regulated the mRNA abundance and protein level of NPC2. Further, NF-κB2 activated NPC2 transcription through direct binding to its promoter region. We also observed cholesterol accumulation in NF-κB2-deficient zebrafish embryo and NF-κB2 mutant mice. Collectively, these data identify a regulatory role for the non-canonical NF-κB pathway in intracellular cholesterol trafficking and suggest a link between cholesterol transport and immune system.