MilliporeSigma
A myosin II nanomachine mimicking the striated muscle.
Nature communications (2018-09-01)
Irene Pertici, Lorenzo Bongini, Luca Melli, Giulio Bianchi, Luca Salvi, Giulia Falorsi, Caterina Squarci, Tamás Bozó, Dan Cojoc, Miklós S Z Kellermayer, Vincenzo Lombardi, Pasquale Bianco
ABSTRACT
The contraction of striated muscle (skeletal and cardiac muscle) is generated by ATP-dependent interactions between the molecular motor myosin II and the actin filament. The myosin motors are mechanically coupled along the thick filament in a geometry not achievable by single-molecule experiments. Here we show that a synthetic one-dimensional nanomachine, comprising fewer than ten myosin II dimers purified from rabbit psoas, performs isometric and isotonic contractions at 2 mM ATP, delivering a maximum power of 5 aW. The results are explained with a kinetic model fitted to the performance of mammalian skeletal muscle, showing that the condition for the motor coordination that maximises the efficiency in striated muscle is a minimum of 32 myosin heads sharing a common mechanical ground. The nanomachine offers a powerful tool for investigating muscle contractile-protein physiology, pathology and pharmacology without the potentially disturbing effects of the cytoskeletal-and regulatory-protein environment.
Sigma-Aldrich
4,4,4-Trifluoro-1-phenyl-1,3-butanedione, 99%