
Zidovudine impurity B

Synonym(s):
3′-Chloro-3′-deoxythymidine
Empirical Formula (Hill Notation):
C10H13ClN2O4
CAS Number:
Molecular Weight:
260.67
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing