Skip to Content
MilliporeSigma
Papanicolaou Stain, Modified EA 65

Papanicolaou Stain, Modified EA 65

Papanicolaou Stain, Modified EA 65

Products

Product Category
Brand
Physical Form
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing