
Moxifloxacin for peak identification A

Synonym(s):
Moxifloxacin, 1-Cyclopropyl-6-fluoro-8-methoxy-7-[(4aS,7aS)-octahydropyrrolo[3,4-b]pyridin-6-yl]-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
Empirical Formula (Hill Notation):
C21H24FN3O4
CAS Number:
Molecular Weight:
401.43
Products
Formula Weight
Manufacturer
Application
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing