
Metoprolol Related Compound B

Synonym(s):
1-Chloro-3-[4-(2-methoxyethyl)phenoxy]- 2-propanol
Empirical Formula (Hill Notation):
C12H17ClO3
CAS Number:
Molecular Weight:
244.71
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing