![Methyl 3-[[(2RS)-2-(dipropylamino)propanoyl]-amino]-4-methylthiophene-2-carboxylate hydrochloride certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/386/957/d524e439-2ea8-42c4-a86f-ffa3878e9091/640/d524e439-2ea8-42c4-a86f-ffa3878e9091.png)
Methyl 3-[[(2RS)-2-(dipropylamino)propanoyl]-amino]-4-methylthiophene-2-carboxylate hydrochloride
![Methyl 3-[[(2RS)-2-(dipropylamino)propanoyl]-amino]-4-methylthiophene-2-carboxylate hydrochloride certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/386/957/d524e439-2ea8-42c4-a86f-ffa3878e9091/640/d524e439-2ea8-42c4-a86f-ffa3878e9091.png)
Synonym(s):
Dipropylarticaine hydrochloride
Empirical Formula (Hill Notation):
C16H26N2O3S · HCl
CAS Number:
Molecular Weight:
362.92
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Quality Segment
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1