Skip to Content
MilliporeSigma
Intermediates for Further Manufacturing Use (FFMU) - ABO Specificities Anti-B from the clone LB-2, Murine IgM, Intermediate for further manufacturing use

Intermediates for Further Manufacturing Use (FFMU) - ABO Specificities

Intermediates for Further Manufacturing Use (FFMU) - ABO Specificities Anti-B from the clone LB-2, Murine IgM, Intermediate for further manufacturing use

Products

Brand
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Quality Segment
Package Size
Technique
Showing 1-8 of 8