
Ethyl indole-3-carboxylate

Synonym(s):
3-(Ethoxycarbonyl)indole, Indole-3-carboxylic acid ethyl ester, NSC 63796
Empirical Formula (Hill Notation):
C11H11NO2
CAS Number:
Molecular Weight:
189.21
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing