
Digitoxin for LC assay

Synonym(s):
Digitoxin, 5β,20(22)-Cardenolide-3β,14-diol-3-(O-2,6-dideoxy-β-D-ribo-hexopyranosyl-[1→4]-O-2,6-dideoxy-β-D-ribo-hexopyranosyl-[1→4]-2,6-dideoxy-β-D-ribo-hexopyranosyl)oxy, Digitoxoside, Lanatoxin
Empirical Formula (Hill Notation):
C41H64O13
CAS Number:
Molecular Weight:
764.94
Products
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Format
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing