
Cefepime dihydrochloride monohydrate for system suitability

Synonym(s):
Cefepime dihydrochloride monohydrate
Empirical Formula (Hill Notation):
C19H25ClN6O5S2 · HCl · H2O
CAS Number:
Molecular Weight:
571.50
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing