Skip to Content
MilliporeSigma
vibrant-m

BRIP human cell line

vibrant-m

Products

Product Category
Biological Source
Tissue
Package Size
Taxon
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing