
Anti-SERPINB6 antibody produced in rabbit

Synonym(s):
Anti-CAP antibody produced in rabbit, Anti-Cytoplasmic antiproteinase antibody produced in rabbit, Anti-PI-6 antibody produced in rabbit, Anti-Placental thrombin inhibitor antibody produced in rabbit, Anti-Protease inhibitor 6 antibody produced in rabbit, Anti-Serpin B6 antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing