
Anti-DDX3 antibody produced in rabbit

Synonym(s):
Anti-DBX, Anti-Dead box polypeptide 3 X-linked, Anti-Helicase-like protein 2, D1PAS1 related sequence 2, D1PAS1-RS2, DBX, DDX3X, DEAD-box RNA helicase DEAD3
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing