
(3-phenylpropyl)methylamine

Synonym(s):
N-Methyl-N-(3-phenylpropyl)amine solution
Empirical Formula (Hill Notation):
C10H15N
CAS Number:
Molecular Weight:
149.23
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing