
1,4-Diethoxy-2-nitro-benzene

Synonym(s):
2,5-Diethoxynitrobenzene
Linear Formula:
(C2H5O)2C6H3NO2
CAS Number:
Molecular Weight:
211.21
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing