Product Name
Monoclonal Anti-PCDHAC2 antibody produced in mouse clone 3D12, purified immunoglobulin, buffered aqueous solution
Product Number
WH0056134M3
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
ELISA (Recombinant protein) | Antibody concentration: 1 ug/ml using recombinant protein. |
Western Blotting (Recombinant protein) | Antibody concentration: 1 ug/ml. Specific band of ~38 kDa using immunogen protein lysate. |