Product Name
Monoclonal Anti-RGS2 antibody produced in mouse clone 4C4, purified immunoglobulin, buffered aqueous solution
Product Number
WH0005997M1
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Western Blotting (Cell lysate) | Antibody concentration: 1 ~ 5 ug/ml. Specific band of ~24 kDa using MCF-7 lysate. |
Immunohistochemistry-Paraffin Embedded Tissue (IHC-P) | Antibody concentration: 3 ug/ml using human tonsil tissue. |
Sandwich ELISA | Detection limit for recombinant GST tagged RGS2 is approximately 0.1ng/ml. |
ELISA (Recombinant protein) | Antibody concentration: 1 ug/ml using recombinant protein. |
Western Blotting (Recombinant protein) | Antibody concentration: 1 ug/ml. Specific band of ~34 kDa using immunogen protein lysate. |