Product Name
Monoclonal Anti-PSMD7 antibody produced in mouse clone 2G5, purified immunoglobulin, buffered aqueous solution
Product Number
WH0005713M1
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Western Blotting (Cell lysate) | Antibody concentration: 1 ~ 5 ug/ml. Specific band of ~37 kDa using Jurkat lysate. |
ELISA (Recombinant protein) | Antibody concentration: 1 ug/ml using recombinant protein. |
Western Blotting (Recombinant protein) | Antibody concentration: 1 ug/ml. Specific band of ~62 kDa using immunogen protein lysate. |