Product Name
Monoclonal Anti-TNFSF8 antibody produced in mouse clone 2E11, ascites fluid
Product Number
WH0000944M1
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
ELISA (Recombinant protein) | Antibody concentration: 1 ug/ml using recombinant protein. |
Western Blotting (Recombinant protein) | Antibody concentration: 1 ug/ml. Specific band of ~35 kDa using immunogen protein lysate. |