Product Name
Monoclonal Anti-CAPS antibody produced in mouse clone 4C6, purified immunoglobulin, buffered aqueous solution
Product Number
WH0000828M1
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Sandwich ELISA | Detection limit for recombinant GST tagged CAPS is approximately 0.03ng/ml. |
ELISA (Recombinant protein) | Antibody concentration: 1 ug/ml using recombinant protein. |
Western Blotting (Recombinant protein) | Antibody concentration: 1 ug/ml. Specific band of ~38 kDa using immunogen protein lysate. |