Product Name
HDAC2, active, GST tagged human recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Product Number
SRP5266
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Purity by SDS-PAGE | >= 70% |
Specific Activity | 3553-5520 RLU/min/ng |