Product Name
HDAC2 FLAG tag human recombinant, expressed in baculovirus infected insect cells, ≥50% (SDS-PAGE)
Product Number
SRP0103
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Concentration | >0.02mg/mL |
Purity (SDS-PAGE) | >=50% |
Specific Activity | >=450 pmol/min/ug. |