Product Name
Anti-MCM7 antibody produced in rabbit
Product Number
SAB5700956
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Concentration | Report result |
Western Blotting | 1:500 - 1:2000 |
ImmunohistochemLstry | 1:50 - 1:200 |
Immunofluorescence | 1:50 - 1:200 |
Immunoprecepitation | 1:50 - 1:200 |