Product Name
Anti-phospho-MAPT (pSer396) antibody produced in rabbit affinity isolated antibody
Product Number
SAB4300081
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance | colorless liquid |
Concentration | 1.0 mg/ml |
ELISA | Not Determined |
Western Blotting | 1:500-1:1000 |
Immunohistochemistry | 1:50-1:100 |
Immunofluorescence | Not Determined |
Immunoprecipitation | Not Determined |
Flow Cytometry | Not Determined |
Chromatin Immunoprecipitation | Not Determined |