Product Name
Monoclonal Anti-LSM4 antibody produced in mouse clone 8A10, purified immunoglobulin
Product Number
SAB1412822
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Sandwich ELISA | Detection limit for recombinant GST tagged LSM4 is 0.1 ng/ml. |
ELISA (Recombinant protein) | Antibody concentration: 1 ug/ml using recombinant protein. |
Western Blotting (Recombinant protein) | Antibody concentration: 1 ug/ml. Specific band of ~33.8 kDa using immunogen protein lysate. |