Product Name
Anti-ANKFY1 antibody produced in mouse purified immunoglobulin, buffered aqueous solution
Product Number
SAB1401696
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Immunofluorescence (IF) | Antibody concentration: 10 ug/ml using HeLa cell. |
Western Blotting (Transfected lysate) | Antibody concentration: 0.1 - 5 ug/ml. Specific band of ~128.7 kDa using ANKFY1 transfected 293T lysate. |
Detection Antibody | Suitable as a detection antibody in sandwich ELISA assay. |