Product Name
Anti-SMCY (C-term) antibody produced in rabbit saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution
Product Number
SAB1300510
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance | colorless liquid |
Concentration | >= 0.25 mg/ml |
ELISA | 1:1000 |
Western Blotting | Not Verified |
Immunohistochemistry | 1:50-1:100 |
Immunofluorescence | Not Verified |
Immunoprecipitation | Not Verified |
Flow Cytometry | Not Verified |
Chromatin Immunoprecipitation | Not Verified |