Product Name
MISSION® esiRNA targeting mouse Plk1
Product Number
EMU039131
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance | colorless liquid |
Concentration | 200 ng/ul |
PCR Product - Gel Electrophoresis | pass |
PCR Product - DNA Sequencing | pass |
Concentration - OD260 or Fluorescence Intensity | pass |
Purified RNaseIII - Gel Electrophoresis | pass |