Product Name
Anti-XTP3TPA (AB1) antibody produced in rabbit affinity isolated antibody
Product Number
AV46329
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
APPEARANCE | WHITE TO OFF-WHITE POWDER |
Western Blotting(Cell lysate) | Suggested dilution at 0.5ug/ml in 5% skim milk / PBS buffer. |