Product Name
Anti-ENO1 (AB1) antibody produced in rabbit IgG fraction of antiserum
Product Number
AV34376
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
APPEARANCE | WHITE TO OFF-WHITE POWDER |
Western Blotting(Cell lysate) | Suggested dilution at 0.5ug/ml in 5% skim milk / PBS buffer. |