Product Name
2-Keto-D-glucose ≥98.0% (TLC)
Product Number
61793
Product Brand
SIGMA
CAS Number
1854-25-7
Molecular Weight
178.14
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Beige |
APPEARANCE (FORM) | Solid |
PURITY (TLC AREA %) | ≥ 98.0 % |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |