Product Name
Minimum Essential Medium Joklik’s Modified with Earle′s Balanced Salts, with 2.0 mM L-glutamine, without calcium chloride, without sodium bicarbonate, dry powder, suitable for cell culture
Product Number
56449C
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance (Colour) | Off-White |
Appearance (Form) | Free Flowing Powder |
pH (w/o NaHCO3) | 5.4 - 5.8 |
pH (w/ 2.0 g/L NaHCO3) | 6.7 - 7.1 |
Osmolality (w/o NaHCO3) | 230 - 270 mOs/kg |
Osmolality (w/ 2.0 g/L NaHCO3) | 270 - 310 mOs/kg |
Solubility | Soluble |
Endotoxin | <= 1.0 EU/ml |
Bioburden (CFU/100mL) | <= 100 |